N-Boc-ethylamine, 97%, Thermo Scientific Chemicals
N-Boc-ethylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-ethylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H32575.06
also known as H32575-06
Price (USD)/ Each
237.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
237.00
Add to cart
N-Boc-ethylamine, 97%, Thermo Scientific Chemicals
Catalog numberH32575.06
Price (USD)/ Each
237.00
-
Add to cart
Chemical Identifiers
CAS38267-76-4
IUPAC Nametert-butyl N-ethylcarbamate
Molecular FormulaC7H15NO2
InChI KeyISTGQSQWSKCNFJ-UHFFFAOYSA-N
SMILESCCNC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystalline or fused solid or lumps or powder
Melting Point (clear melt)40-45?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only