Thermo Scientific Chemicals

3-Chloro-4-nitroaniline, 95%, Thermo Scientific Chemicals

Catalog number: H32581.03
1 g, Each
Thermo Scientific Chemicals

3-Chloro-4-nitroaniline, 95%, Thermo Scientific Chemicals

Catalog number: H32581.03
1 g, Each
Quantity
Catalog number: H32581.03
also known as H32581-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
825-41-2
IUPAC Name
3-chloro-4-nitroaniline
Molecular Formula
C6H5ClN2O2
InChI Key
LDSIOPGMLLPSSR-UHFFFAOYSA-N
SMILES
NC1=CC=C(C(Cl)=C1)[N+]([O-])=O
Appearance (Color)
Yellow
Assay (GC)
≥94.0%
Melting Point (clear melt)
153.0-165.0?C
Form
Powder

Description

3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text