3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Chloro-4-nitroaniline is used as a pharmaceutical and medical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
M.M.V. Ramana.; S.S. Malik.; J.A. Parihar. Guanidinium nitrate: a novel reagent for aryl nitrations. Tetrahedron Letters. 2004, 45 (47), 8681-8683.
W. Greizerstein.; R. A. Bonelli.; J. A. Brieux. Polar Effects of Substitutents on the Reaction Rates of 4-R- and 5-R-2-Nitrochlorobenzenes with Piperidine in Benzene. J. Am. Chem. Soc. 1962, 84 (6), 1026-1032.