Thermo Scientific Chemicals

Ammonium O,O'-dimethyldithiophosphate, 95%, Thermo Scientific Chemicals

Catalog number: H32592.03
1 g, Each
Thermo Scientific Chemicals

Ammonium O,O'-dimethyldithiophosphate, 95%, Thermo Scientific Chemicals

Catalog number: H32592.03
1 g, Each
Quantity
Catalog number: H32592.03
also known as H32592-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1066-97-3
IUPAC Name
ammonium O,O-dimethyl sulfanidylphosphonothioate
Molecular Formula
C2H10NO2PS2
InChI Key
PPGORMGERPBFTJ-UHFFFAOYSA-N
SMILES
[NH4+].COP([S-])(=S)OC
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥94.0 to ≤106.0%
Melting Point (clear melt)
138.0-150.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text