4-Iodo-1-phenyl-1H-pyrazole, 95%, Thermo Scientific Chemicals
4-Iodo-1-phenyl-1H-pyrazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodo-1-phenyl-1H-pyrazole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32612.MD
also known as H32612-MD
Price (USD)/ Each
149.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
149.00
Add to cart
4-Iodo-1-phenyl-1H-pyrazole, 95%, Thermo Scientific Chemicals
Catalog numberH32612.MD
Price (USD)/ Each
149.00
-
Add to cart
Chemical Identifiers
CAS23889-85-2
IUPAC Name4-iodo-1-phenyl-1H-pyrazole
Molecular FormulaC9H7IN2
InChI KeyPYJVDCYRDQUUKO-UHFFFAOYSA-N
SMILESIC1=CN(N=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granular solid
Assay (GC)≥94.0%
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only