2,4-Difluoro-5-nitrobenzoic acid, 97%, Thermo Scientific Chemicals
2,4-Difluoro-5-nitrobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Difluoro-5-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32617.06
also known as H32617-06
Price (USD)/ Each
124.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
124.00
Add to cart
2,4-Difluoro-5-nitrobenzoic acid, 97%, Thermo Scientific Chemicals
Catalog numberH32617.06
Price (USD)/ Each
124.00
-
Add to cart
Chemical Identifiers
CAS153775-33-8
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)121.0-130.0?C
Appearance (Color)White to cream to pale yellow
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only