Thermo Scientific Chemicals

Ethyl 1-Boc-DL-nipecotate, 97%, Thermo Scientific Chemicals

Catalog number: H32704.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 1-Boc-DL-nipecotate, 97%, Thermo Scientific Chemicals

Catalog number: H32704.06
5 g, Each
Quantity
Catalog number: H32704.06
also known as H32704-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
130250-54-3
IUPAC Name
1-tert-butyl 3-ethyl piperidine-1,3-dicarboxylate
Molecular Formula
C13H23NO4
InChI Key
YCXCRFGBFZTUSU-UHFFFAOYNA-N
SMILES
CCOC(=O)C1CCCN(C1)C(=O)OC(C)(C)C
Appearance (Color)
White
Form
Fused solid
Assay (GC)
≥96.0%
Melting Point (clear melt)
30.0-39.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text