N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Hyeung-geun Park.; Byeong-seon Jeong.; Mi-sook Yoo.; Mi-kyoung Park.; Hoon Huh.; Sang-sup Jew. Trimeric Cinchona alkaloid phase-transfer catalyst: α,α',α″-tris[O(9)-allylcinchonidinium]mesitylene tribromide. Tetrahedron Letters. 2001, 42 (28), 4645-4648.
- Keiji Maruoka.; Takashi Ooi. Enantioselective Amino Acid Synthesis by Chiral Phase-Transfer Catalysis. Chem. Rev. 2003, 103 (8), 3013-3028.