Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, 95%, Thermo Scientific Chemicals
Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H32731.03
also known as H32731-03
Price (USD)/ Each
203.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
203.00
Add to cart
Methyl 4-(hydroxymethyl)pyridine-2-carboxylate, 95%, Thermo Scientific Chemicals
Catalog numberH32731.03
Price (USD)/ Each
203.00
-
Add to cart
Chemical Identifiers
CAS317335-15-2
IUPAC Namemethyl 4-(hydroxymethyl)pyridine-2-carboxylate
Molecular FormulaC8H9NO3
InChI KeyWEIFQBALQLWZFZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=NC=CC(CO)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Non-aqueous acid-base Titration)≥94.0 to ≤106.0%
Melting Point (clear melt)74-83?C
Assay (Silylated GC)≥94.0%
Appearance (Color)White to pale yellow
Methyl 4-(hydroxymethyl)pyridine-2 is used as an intermediate in pharmaceuticals and also finds use in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-(hydroxymethyl)pyridine-2 is used as an intermediate in pharmaceuticals and also finds use in chemical research.

Notes
Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
RUO – Research Use Only