2-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals
2-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H32737.06
also known as H32737-06
Price (USD)/ Each
572.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
572.00
Add to cart
2-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals
Catalog numberH32737.06
Price (USD)/ Each
572.00
-
Add to cart
Chemical Identifiers
CAS42590-97-6
IUPAC Name2-methoxybenzene-1-carbothioamide
Molecular FormulaC8H9NOS
InChI KeyYNEHBLLUZJTDOL-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)110.5-119.5?C
Appearance (Color)Yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only