Thermo Scientific Chemicals

Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H32755.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H32755.06
5 g, Each
Quantity
Catalog number: H32755.06
also known as H32755-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108354-78-5
IUPAC Name
methyl 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate
Molecular Formula
C10H13NO2S
InChI Key
DKYYKIHEIOOWRB-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(N)SC2=C1CCCC2
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Appearance (Color)
Pale cream to cream to yellow
Assay (GC)
≥97.5%
Melting Point (clear melt)
128.0-134.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text