4-Hydroxythiobenzamide, 98%, Thermo Scientific Chemicals
4-Hydroxythiobenzamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxythiobenzamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32780.06
also known as H32780-06
Price (USD)/ Each
114.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
114.00
Add to cart
4-Hydroxythiobenzamide, 98%, Thermo Scientific Chemicals
Catalog numberH32780.06
Price (USD)/ Each
114.00
-
Add to cart
Chemical Identifiers
CAS25984-63-8
IUPAC Name4-[amino(sulfanyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular FormulaC7H7NOS
InChI KeyDOCQBKMMNPJLOR-UHFFFAOYSA-N
SMILESNC(S)=C1C=CC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormPowder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only