6-Fluorochromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
6-Fluorochromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Fluorochromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Catalog number H32857.03
also known as H32857-03
Price (USD)/ Each
28.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
28.10
Add to cart
6-Fluorochromone-3-carboxaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH32857.03
Price (USD)/ Each
28.10
-
Add to cart
Chemical Identifiers
CAS69155-76-6
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)152.0-161.0?C
Appearance (Color)Pale yellow
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only