Thermo Scientific Chemicals

3-(Trifluoromethyl)salicylaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: H32858.03
1 g, Each
Thermo Scientific Chemicals

3-(Trifluoromethyl)salicylaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: H32858.03
1 g, Each
Quantity
Catalog number: H32858.03
also known as H32858-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
336628-67-2
Appearance (Color)
White to yellow or grey
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%
Melting Point
53-60?C
Form
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text