Thermo Scientific Chemicals

3-Benzyloxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32882.03
1 g, Each
Thermo Scientific Chemicals

3-Benzyloxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32882.03
1 g, Each
Quantity
Catalog number: H32882.03
also known as H32882-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69026-14-8
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Melting Point (clear melt)
132.0-138.0?C
Form
Crystals or powder or crystalline powder
Assay (Silylated GC)
≥97.5%

Description

3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzyloxybenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text