Thermo Scientific Chemicals

4-Acetylbenzenesulfonamide, 97%, Thermo Scientific Chemicals

Catalog number: H32901.06
5 g, Each
Thermo Scientific Chemicals

4-Acetylbenzenesulfonamide, 97%, Thermo Scientific Chemicals

Catalog number: H32901.06
5 g, Each
Quantity
Catalog number: H32901.06
also known as H32901-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1565-17-9
IUPAC Name
4-acetylbenzene-1-sulfonamide
Molecular Formula
C8H9NO3S
InChI Key
CSATVXJBGFVJES-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(C=C1)S(N)(=O)=O
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to yellow
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
174.0-183.0?C

Description

4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

Solubility
soluble in water( 2650 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text