Ethyl benzothiazole-2-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl benzothiazole-2-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl benzothiazole-2-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H32904.06
also known as H32904-06
Price (USD)/ Each
70.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
70.20
Add to cart
Ethyl benzothiazole-2-carboxylate, 98%, Thermo Scientific Chemicals
Catalog numberH32904.06
Price (USD)/ Each
70.20
-
Add to cart
Chemical Identifiers
CAS32137-76-1
IUPAC Nameethyl 1,3-benzothiazole-2-carboxylate
Molecular FormulaC10H9NO2S
InChI KeyVLQLCEXNNGQELL-UHFFFAOYSA-N
SMILESCCOC(=O)C1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Pale cream to pale brown
Assay (GC)≥97.5%
Melting Point (clear melt)66.0-73.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only