Thermo Scientific Chemicals

2-Chloro-4-fluoro-1-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: H32906.03
1 g, Each
Thermo Scientific Chemicals

2-Chloro-4-fluoro-1-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: H32906.03
1 g, Each
Quantity
Catalog number: H32906.03
also known as H32906-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2106-50-5
IUPAC Name
2-chloro-4-fluoro-1-nitrobenzene
Molecular Formula
C6H3ClFNO2
InChI Key
KQOOFMWRLDRDAX-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(F)C=C1Cl
Appearance (Color)
Pale yellow to yellow to yellow-brown
Form
Fused solid or lumps or crystals or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
32-37?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text