Thermo Scientific Chemicals

2-Amino-5-(trifluoromethoxy)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32916.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-5-(trifluoromethoxy)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: H32916.06
5 g, Each
Quantity
Catalog number: H32916.06
also known as H32916-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83265-56-9
IUPAC Name
2-amino-5-(trifluoromethoxy)benzoic acid
Molecular Formula
C8H6F3NO3
InChI Key
UXNGDCBPIGOZFO-UHFFFAOYSA-N
SMILES
NC1=C(C=C(OC(F)(F)F)C=C1)C(O)=O
Assay (Silylated GC)
≥97.5%
Melting Point
138.0-144.0?C
Appearance (Color)
White to cream or pale yellow
Assay (Aqueous acid-base Titration)
≥97.5% to ≤102.5%
Form
Crystals or powder or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text