2-Methylbenzothiazol-5-ol, 97%, Thermo Scientific Chemicals
2-Methylbenzothiazol-5-ol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methylbenzothiazol-5-ol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H32937.03
also known as H32937-03
Price (USD)/ Each
40.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
40.00
Add to cart
2-Methylbenzothiazol-5-ol, 97%, Thermo Scientific Chemicals
Catalog numberH32937.03
Price (USD)/ Each
40.00
-
Add to cart
Chemical Identifiers
CAS68867-14-1
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Anke Spurg.; Siegfried R. Waldvogel. High-Yielding Cleavage of (Aryloxy)acetates. European Journal of Organic Chemistry. 2008, 2008 (2), 337-342.
  2. Janina Kabatc.; Jerzy Paczkowski. The photophysical and photochemical properties of the oxacarbocyanine and thiacarbocyanine dyes. Dyes and Pigments. 2004, 61 (1), 1-16.