Thermo Scientific Chemicals

3-Bromobenzo[b]thiophene, 95%, Thermo Scientific Chemicals

Catalog number: H32948.18
50 g, Each
Thermo Scientific Chemicals

3-Bromobenzo[b]thiophene, 95%, Thermo Scientific Chemicals

Catalog number: H32948.18
50 g, Each
Quantity
Catalog number: H32948.18
also known as H32948-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7342-82-7
IUPAC Name
3-bromo-1-benzothiophene
Molecular Formula
C8H5BrS
InChI Key
SRWDQSRTOOMPMO-UHFFFAOYSA-N
SMILES
BrC1=CSC2=CC=CC=C12
Refractive Index
1.6640-1.6700 @ 20?C
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow to red
Form
Liquid
Assay (GC)
≥94.0%

Description

Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.

Solubility
Sparingly Soluble (0.14 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text