Thermo Scientific Chemicals

4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: H32982.06
5 g, Each
Thermo Scientific Chemicals

4-Bromo-2,3,5,6-tetrafluorobenzoic acid, 97+%, Thermo Scientific Chemicals

Catalog number: H32982.06
5 g, Each
Quantity
Catalog number: H32982.06
also known as H32982-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4707-24-8
IUPAC Name
4-bromo-2,3,5,6-tetrafluorobenzoic acid
Molecular Formula
C7HBrF4O2
InChI Key
MTZLICDXLGQWJV-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(F)C(F)=C(Br)C(F)=C1F
Assay (Silylated GC)
>97.0%
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
>97.0% (dry wt. basis)
Loss on Drying
<6.0%
Form
Powder

Description

4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,3,5,6-tetrafluorobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text