Thermo Scientific Chemicals

(4-Imidazolyl)acetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H32992.03
1 g, Each
Thermo Scientific Chemicals

(4-Imidazolyl)acetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: H32992.03
1 g, Each
Quantity
Catalog number: H32992.03
also known as H32992-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18502-05-1
IUPAC Name
2-(1H-imidazol-5-yl)acetonitrile
Molecular Formula
C5H5N3
InChI Key
DQZBHUXBFNBJLX-UHFFFAOYSA-N
SMILES
N#CCC1=CN=CN1
Melting Point (clear melt)
129.5-138.5?C
Appearance (Color)
Orange to brown
Assay (GC)
≥96.0%
Form
Powder

Description

(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text