Thermo Scientific Chemicals

3-Cyano-5-hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: H33006.MD
250 mg, Each
Thermo Scientific Chemicals

3-Cyano-5-hydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: H33006.MD
250 mg, Each
Quantity
Catalog number: H33006.MD
also known as H33006-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
152803-24-2
Form
Powder
Appearance (Color)
Pale cream to pale brown
Assay (HPLC)
≥96.0%

Description

3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text