2,2'-O-Cyclouridine, 98%, Thermo Scientific Chemicals
2,2'-O-Cyclouridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2'-O-Cyclouridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33038.03
also known as H33038-03
Price (USD)/ Each
32.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
32.00
Add to cart
2,2'-O-Cyclouridine, 98%, Thermo Scientific Chemicals
Catalog numberH33038.03
Price (USD)/ Each
32.00
-
Add to cart
Chemical Identifiers
CAS3736-77-4
IUPAC Name(2R,4R,5R,6S)-5-hydroxy-4-(hydroxymethyl)-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-10-one
Molecular FormulaC9H10N2O5
InChI KeyUUGITDASWNOAGG-CCXZUQQUSA-N
SMILESOC[C@H]1O[C@@H]2[C@@H](OC3=NC(=O)C=CN23)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)242.0-249.0?C
FormPowder
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only