Thermo Scientific Chemicals

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33055.MD
250 mg, Each
Thermo Scientific Chemicals

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33055.MD
250 mg, Each
Quantity
Catalog number: H33055.MD
also known as H33055-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
128073-33-6
IUPAC Name
2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
Molecular Formula
C5H6F2O2
InChI Key
HLFLYOQLHYYNLT-UHFFFAOYNA-N
SMILES
CC1(CC1(F)F)C(O)=O
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
White to yellow to pale brown
Melting Point (clear melt)
60-66?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text