Thermo Scientific Chemicals

3-(4-Chloro-3-methylphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33072.ME
500 mg, Each
Thermo Scientific Chemicals

3-(4-Chloro-3-methylphenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33072.ME
500 mg, Each
Quantity
Catalog number: H33072.ME
also known as H33072-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1086386-05-1
IUPAC Name
3-(4-chloro-3-methylphenyl)propanoic acid
Molecular Formula
C10H11ClO2
InChI Key
PCLIPGMIRVJYFB-UHFFFAOYSA-N
SMILES
CC1=C(Cl)C=CC(CCC(O)=O)=C1
Form
Powder
Assay (Silylated GC)
≥95.0%
Appearance (Color)
White
Melting Point (clear melt)
85.0-94.0?C
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text