3,3'-Diethyloxadicarbocyanine iodide, 96%, Thermo Scientific Chemicals
3,3'-Diethyloxadicarbocyanine iodide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3'-Diethyloxadicarbocyanine iodide, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H33094.06
also known as H33094-06
Price (USD)/ Each
386.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
386.00
Add to cart
3,3'-Diethyloxadicarbocyanine iodide, 96%, Thermo Scientific Chemicals
Catalog numberH33094.06
Price (USD)/ Each
386.00
-
Add to cart
Chemical Identifiers
CAS14806-50-9
SpecificationsSpecification SheetSpecification Sheet
Assay (special)≥95.0 to ≤105.0% (based on elemental I and N)
Elemental AnalysisN: 5.47-6.05% (Theory: 5.76%)
Appearance (Color)Dark blue
Elemental AnalysisI: 24.79-27.40% (Theory: 26.09%)
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only