Thermo Scientific Chemicals

2,1,3-Benzothiadiazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33095.03
1 g, Each
Thermo Scientific Chemicals

2,1,3-Benzothiadiazole-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H33095.03
1 g, Each
Quantity
Catalog number: H33095.03
also known as H33095-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16405-98-4
IUPAC Name
2,1,3-benzothiadiazole-5-carboxylic acid
Molecular Formula
C7H4N2O2S
InChI Key
YHMXJZVGBCACMT-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC2=NSN=C2C=C1
Form
Powder
Appearance (Color)
Pale cream
Assay (Silylated GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
226.5-232.5?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text