Thermo Scientific Chemicals

1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals

Catalog number: H33116.03
1 g, Each
Thermo Scientific Chemicals

1,3,5-Trifluoro-2,4,6-triiodobenzene, 97%, Thermo Scientific Chemicals

Catalog number: H33116.03
1 g, Each
Quantity
Catalog number: H33116.03
also known as H33116-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84322-56-5
IUPAC Name
1,3,5-trifluoro-2,4,6-triiodobenzene
Molecular Formula
C6F3I3
InChI Key
NTAZOPPTLZSXQH-UHFFFAOYSA-N
SMILES
FC1=C(I)C(F)=C(I)C(F)=C1I
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream to cream to brown to gray
Melting Point (clear melt)
151.0-160.0?C

Description

1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text