1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Trifluoro-2,4,6-triiodobenzene is useful for biological applications. Further, it is used as an intermediate in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only
Lieffrig, J.; Jeannin, O.; Fourmigué, M. Expanded Halogen-Bonded Anion Organic Networks with Star-Shaped Iodoethynyl-Substituted Molecules: From Corrugated 2D Hexagonal Lattices to Pyrite-Type 2-Fold Interpenetrated Cubic Lattices. J. Am. Chem. Soc. 2013, 135 (16), 6200-6210.
Ji, B.; Zhang, Y.; Deng, D.; Wang, W. Improper halogen bond in the crystal structure. CrystEngComm 2013, 15 (16), 3093-3096.