Thermo Scientific Chemicals

6-Chloro-2-iodo-3-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: H33127.03
1 g, Each
Thermo Scientific Chemicals

6-Chloro-2-iodo-3-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: H33127.03
1 g, Each
Quantity
Catalog number: H33127.03
also known as H33127-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
188057-26-3
IUPAC Name
6-chloro-2-iodopyridin-3-ol
Molecular Formula
C5H3ClINO
InChI Key
FWIMPBYPMQSSCD-UHFFFAOYSA-N
SMILES
OC1=C(I)N=C(Cl)C=C1
Appearance (Color)
White to pale cream
Form
Powder
Assay (Silylated GC)
≥97.5%
Melting Point (clear melt)
188.0-194.0?C

Description

2,2'-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.

Solubility
Slightly Soluble (1.3 g/L) (25°C).

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text