2,2'-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2′-Dipyridylamine has been obtained by by heating a mixture of 2-aminopyridine and 2-bromo- (chloro-, iodo- ) -pyridine with anhydrous zinc chloride or with barium oxide.
Solubility
Slightly Soluble (1.3 g/L) (25°C).
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
RUO – Research Use Only
C. Zwart ,;J. P. Wibaut. Synthesis of four dipyridylamines. Recueil des Travaux Chimiques des Pays-Bas. 1955, 74 (9) ,1081-1084.
authors. title . Journal name. year of publication , volume issue , pg no.