Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 g
5000 g
Catalog number H33152.0B
also known as H33152-0B
Price (USD)/ Each
51.65
Online exclusive
56.70 
Save 5.05 (9%)
-
Add to cart
Quantity:
1000 g
Request bulk or custom format
Price (USD)/ Each
51.65
Online exclusive
56.70 
Save 5.05 (9%)
Add to cart
Filter aid, Celite∣r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
Catalog numberH33152.0B
Price (USD)/ Each
51.65
Online exclusive
56.70 
Save 5.05 (9%)
-
Add to cart
Chemical Identifiers
CAS68855-54-9
SpecificationsSpecification SheetSpecification Sheet
Permeability2.60-6.50 darcy
Appearance (Color)White to cream to grey
CommentMay contain a small quantity of dark particulates (grit) from the manufacturing process.
Wet density (Centrifuged)255-415 g/L
FormPowder
View more
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
RUO – Research Use Only

General References:

  1. J A Zeevaart. Effects of photoperiod on growth rate and endogenous gibberellins in the long-day rosette plant spinach. Plant Physiology. 197, 1971(6), 821-827.
  2. N CITRI et. al.The effect of urea and guanidine hydrochloride on activity and optical rotation of penicillinase.. Journal of Biological Chemistry. 1960, 235 3454-3459.