Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
RUO – Research Use Only
General References:
- J A Zeevaart. Effects of photoperiod on growth rate and endogenous gibberellins in the long-day rosette plant spinach. Plant Physiology. 197, 1971(6), 821-827.
- N CITRI et. al.The effect of urea and guanidine hydrochloride on activity and optical rotation of penicillinase.. Journal of Biological Chemistry. 1960, 235 3454-3459.