5-Fluorobenzo[b]thiophene-2-carboxylic acid, 96%, Thermo Scientific Chemicals
5-Fluorobenzo[b]thiophene-2-carboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Fluorobenzo[b]thiophene-2-carboxylic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33239.03
also known as H33239-03
Price (USD)/ Each
74.30
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
74.30
Add to cart
5-Fluorobenzo[b]thiophene-2-carboxylic acid, 96%, Thermo Scientific Chemicals
Catalog numberH33239.03
Price (USD)/ Each
74.30
-
Add to cart
Chemical Identifiers
CAS70060-13-8
IUPAC Name5-fluoro-1-benzothiophene-2-carboxylate
Molecular FormulaC9H4FO2S
InChI KeyPLVPMOSTGNZKQQ-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC2=CC(F)=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
FormPowder
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (HPLC)≥95.0%
Melting Point (clear melt)257.0-266.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only