Thermo Scientific Chemicals

4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Catalog number: H33260.06
5 g, Each
Thermo Scientific Chemicals

4-Chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Catalog number: H33260.06
5 g, Each
Quantity
Catalog number: H33260.06
also known as H33260-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
331761-46-7
IUPAC Name
4-chloro-5-(4-chlorophenyl)thieno[2,3-d]pyrimidine
Molecular Formula
C12H6Cl2N2S
InChI Key
QDMQHIKVIOWCGY-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C1)C1=CSC2=NC=NC(Cl)=C12
Appearance (Color)
White to pale green or pale brown
Form
Powder
Assay (GC)
≥95.0%
Melting Point
139.0-148.0°C
Proton NMR
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text