Thermo Scientific Chemicals

3-(3-Chlorophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33266.06
5 g, Each
Thermo Scientific Chemicals

3-(3-Chlorophenyl)propionic acid, 96%, Thermo Scientific Chemicals

Catalog number: H33266.06
5 g, Each
Quantity
Catalog number: H33266.06
also known as H33266-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21640-48-2
Assay (Aqueous acid-base Titration)
≥95.0 to ≤105.0%
Form
Powder
Melting Point (clear melt)
66.5-75.5?C
Appearance (Color)
White
Assay (HPLC)
≥95.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text