Thermo Scientific Chemicals

Di-2-thienyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: H33284.03
1 g, Each
Thermo Scientific Chemicals

Di-2-thienyl sulfide, 97%, Thermo Scientific Chemicals

Catalog number: H33284.03
1 g, Each
Quantity
Catalog number: H33284.03
also known as H33284-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3988-99-6
IUPAC Name
2-(thiophen-2-ylsulfanyl)thiophene
Molecular Formula
C8H6S3
InChI Key
AKYIWBKPINOXJY-UHFFFAOYSA-N
SMILES
S(C1=CC=CS1)C1=CC=CS1
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Refractive Index
1.6595-1.6645 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text