Di-2-thienyl sulfide, 97%, Thermo Scientific Chemicals
Di-2-thienyl sulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-2-thienyl sulfide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H33284.06
also known as H33284-06
Price (USD)/ Each
227.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
227.00
Add to cart
Di-2-thienyl sulfide, 97%, Thermo Scientific Chemicals
Catalog numberH33284.06
Price (USD)/ Each
227.00
-
Add to cart
Chemical Identifiers
CAS3988-99-6
IUPAC Name2-(thiophen-2-ylsulfanyl)thiophene
Molecular FormulaC8H6S3
InChI KeyAKYIWBKPINOXJY-UHFFFAOYSA-N
SMILESS(C1=CC=CS1)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow to brown
FormLiquid
Refractive Index1.6595-1.6645 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only