Thermo Scientific Chemicals

5-Aminobenzothiazole, 96%, Thermo Scientific Chemicals

Catalog number: H33309.06
5 g, Each
Thermo Scientific Chemicals

5-Aminobenzothiazole, 96%, Thermo Scientific Chemicals

Catalog number: H33309.06
5 g, Each
Quantity
Catalog number: H33309.06
also known as H33309-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1123-93-9
IUPAC Name
1,3-benzothiazol-5-amine
Molecular Formula
C7H6N2S
InChI Key
UJZYHMZRXGNDFB-UHFFFAOYSA-N
SMILES
NC1=CC=C2SC=NC2=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
Cream to brown
Assay (GC)
≥95.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
68.0-78.0?C

Description

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text