Thermo Scientific Chemicals

Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H33319.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 2-amino-3,3,3-trifluoropropionate hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H33319.03
1 g, Each
Quantity
Catalog number: H33319.03
also known as H33319-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
193140-71-5
IUPAC Name
ethyl 2-amino-3,3,3-trifluoropropanoate hydrochloride
Molecular Formula
C5H9ClF3NO2
InChI Key
MNFISCBCWBUTTQ-UHFFFAOYNA-N
SMILES
Cl.CCOC(=O)C(N)C(F)(F)F
Appearance (Color)
White to pale cream
Assay (Titration ex Chloride)
≥95.0 to ≤105.0%
Form
Powder
Water Content (Karl Fischer Titration)
≤2%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text