Coumarin 334, 96%, Thermo Scientific Chemicals
Coumarin 334, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Coumarin 334, 96%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
500 mg
Catalog number H33332.ME
also known as H33332-ME
Price (USD)/ Each
72.65
Online exclusive
80.30 
Save 7.65 (10%)
-
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.30 
Save 7.65 (10%)
Add to cart
Coumarin 334, 96%, Thermo Scientific Chemicals
Catalog numberH33332.ME
Price (USD)/ Each
72.65
Online exclusive
80.30 
Save 7.65 (10%)
-
Add to cart
Chemical Identifiers
CAS55804-67-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to dark orange or red
Assay (HPLC)≥95.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)176.0-186.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only