trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H33367.03
also known as H33367-03
Price (USD)/ Each
177.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
177.00
Add to cart
trans-2-Ethoxyethenyl-1-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog numberH33367.03
Price (USD)/ Each
177.00
-
Add to cart
Chemical Identifiers
CAS1201905-61-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Refractive Index1.4425-1.4485 @ 20?C
Assay (GC)≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Crestey, F.; Hooyberghs, G.; Kristensen, J. L. Concise synthesis of new bridged-nicotine analogues. Tetrahedron 2012, 68 (5), 1417-1421.
  2. Petersen, I. N.; Crestey, F.; Jensen, A. A.; Indurthi, D. C.; Pedersen, H.; Andreasen, J. T.; Balle, T.; Kristensen, J. L. Tying up Nicotine: New Selective Competitive Antagonist of the Neuronal Nicotinic Acetylcholine Receptors. ACS Med. Chem. Lett. 2015, 6 (4), 472-475.
  3. Innocenti, P.; Woodward, H.; O'Fee, L.; Hoelder, S. Expanding the scope of fused pyrimidines as kinase inhibitor scaffolds: synthesis and modification of pyrido[3,4-d]pyrimidines. Org. Biomol. Chem. 2015, 13 (3), 893-904.