Thermo Scientific Chemicals

Hydrogen chloride, 3M in cyclopentyl methyl ether, Thermo Scientific Chemicals

Catalog number: H33398.AP
500 mL, Each
Thermo Scientific Chemicals

Hydrogen chloride, 3M in cyclopentyl methyl ether, Thermo Scientific Chemicals

Catalog number: H33398.AP
500 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H33398.AP
also known as H33398-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-01-0
IUPAC Name
hydrogen chloride
Molecular Formula
ClH
InChI Key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES
Cl
Appearance (Color)
Clear colorless
Form
Liquid
Assay (Aqueous acid-base Titration)
2.65-3.25M

Description

In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pesticides, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production. CPME has proved to be quite useful as a process solvent for organic synthesis, simply because it is free of the drawbacks present in the more traditional solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text