Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H33407.06
also known as H33407-06
Price (USD)/ Each
383.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
383.00
Add to cart
Methyl 1-Boc-pyrrolidine-3-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberH33407.06
Price (USD)/ Each
383.00
-
Add to cart
Chemical Identifiers
CAS122684-33-7
IUPAC Name1-tert-butyl 3-methyl pyrrolidine-1,3-dicarboxylate
Molecular FormulaC11H19NO4
InChI KeyLIWFYAVKYUQMRE-UHFFFAOYNA-N
SMILESCOC(=O)C1CCN(C1)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4555-1.4605 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only