Thermo Scientific Chemicals

4-(Pentafluorothio)phenol, 97%, Thermo Scientific Chemicals

Catalog number: H33413.06
5 g, Each
Thermo Scientific Chemicals

4-(Pentafluorothio)phenol, 97%, Thermo Scientific Chemicals

Catalog number: H33413.06
5 g, Each
Quantity
Catalog number: H33413.06
also known as H33413-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
774-94-7
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Appearance (Color)
White to cream
Melting Point (clear melt)
105-111°C

Description

Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text