1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H33424.03
also known as H33424-03
Price (USD)/ Each
89.60
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
89.60
Add to cart
1-(4-Trifluoromethyl-2-pyrimidinyl)-1H-pyrazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals
Catalog numberH33424.03
Price (USD)/ Each
89.60
-
Add to cart
Chemical Identifiers
CAS1215564-15-0
IUPAC Name1-[4-(trifluoromethyl)pyrimidin-2-yl]-1H-pyrazole-4-sulfonyl chloride
Molecular FormulaC8H4ClF3N4O2S
InChI KeySADUDBJMXGGXDE-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC(=NC=C1)N1C=C(C=N1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Assay (GC)≥94.0%
Melting Point (clear melt)92.0-105.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only