Thermo Scientific Chemicals
Palladium, 5% on carbon, Type 87L, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: H33434.09
10 g, Each
Thermo Scientific Chemicals
Palladium, 5% on carbon, Type 87L, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: H33434.09
10 g, Each
Quantity
Catalog number: H33434.09
also known as H33434-09
Price (USD)
Have Questions?
Appearance (Color)
Dark grey to black
Elemental Analysis
(Palladium): 4.75-5.25% (dry wt. basis)
Loss on Drying
50.0-65.0%
Form
Powder