Palladium, 5% on carbon, Type 87L, nominally 50% water wet, Thermo Scientific Chemicals
Palladium, 5% on carbon, Type 87L, nominally 50% water wet, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 5% on carbon, Type 87L, nominally 50% water wet, Thermo Scientific Chemicals

 
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number H33434.18
also known as H33434-18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPalladium
Name Note5% on carbon, Type 87L, nominally 50% water wet
CAS7440-05-3
ColorGray
Physical FormPowder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only